p-Toluonitril 98+%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Methylbenzoesäurenitril, 4-Methylbenzonitril, p-Methylbenzonitril
158021000
158022500
158020050
ACRO158021000EA
44.8
EUR
ACRO158021000
ACRO158022500
ACRO158020050
p-Toluonitril 98+%
p-Toluonitril
Formel:
C₈H₇N Molecular Weight: 117,15 g/mol Siedepunkt: 103…106 °C (20 mmHg) Schmelzpunkt: 26…28 °C Dichte: 0,98 g/cm³ Flammpunkt: 85 °C |
MDL:
MFCD00001827 CAS-Nummer: 104-85-8 EINECS: 203-244-8 Merck Index: 12,09676 |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid or solid |
Infrared spectrum | Conforms |
GC | ≥98.5 % |
Water | ≤0.01 % |
UV | at 280 nm A: ≤0.05 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.0005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...