5-Hydroxy-2-indolcarbonsäure 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Hydroxy-2-indolecarboxylic acid, 5-Hydroxy-1H-indole-2-carboxylic acid
162450010
162450050
ACRO162450010EA
62.8
EUR
ACRO162450010
ACRO162450050
5-Hydroxy-2-indolcarbonsäure 98%
5-Hydroxyindole-2-carboxylic acid
Formel:
C₉H₇NO₃ Molecular Weight: 177,16 g/mol Schmelzpunkt: 249 °C (Zersetzung) Lagertemperatur: Kühlschrank |
MDL:
MFCD00005615 CAS-Nummer: 21598-06-1 EINECS: 244-468-6 |
Spezifikation Testergebnisse
Appearance | Light yellow to beige to brown Crystalline powder |
Infrared spectrum | Conforms |
Titration with NaOH | 97.5 to 102.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...