2-Amino-6-methoxybenzothiazol 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 6-Methoxybenzothiazol-2-ylamine

172020500 172020050
ACRO172020500EA 91.5 EUR
ACRO172020500 ACRO172020050
2-Amino-6-methoxybenzothiazol 98%
2-Amino-6-methoxybenzothiazole
Formel: C₈H₈N₂OS
Molecular Weight: 180.23 g/mol
Siedepunkt: 348
Schmelzpunkt: 162…169 °C
MDL: MFCD00005787
CAS-Nummer: 1747-60-0
EINECS: 217-130-0
UN: 2811

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 165 to 169 °C
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR