m-Phenylendiamin Dihydrochlorid 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: m-Phenylendiammoniumdichlorid, 1,3-Phenylendiamin Dihydrochlorid, Benzol-1,3-diamindihydrochlorid, 1,3-Diaminobenzol Dihydrochlorid, 1,3-Phenylendiammoniumdichlorid

216650250 216651000
ACRO216650250EA 47.7 EUR
ACRO216650250 ACRO216651000
m-Phenylendiamin Dihydrochlorid 99%
m-Phenylendiamin Dihydrochlorid
Formel: C₆H₁₀Cl₂N₂
Molecular Weight: 181.06 g/mol
Schmelzpunkt: 277…278 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012975
CAS-Nummer: 541-69-5
EINECS: 208-790-0
UN: 1673
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance Light beige to light grey Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR