N-Phenyl-p-phenylendiamin 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Aminodiphenylamin, N-(4-Aminophenyl)anilin

221000010 221001000 221002500
ACRO221000010EA 271 EUR
ACRO221000010 ACRO221001000 ACRO221002500
N-Phenyl-p-phenylendiamin 98%
N-Phenyl-p-phenylendiamin
Formel: C₁₂H₁₂N₂
Molecular Weight: 184,24 g/mol
Siedepunkt: 354 °C (1013 hPa)
Schmelzpunkt: 69…75 °C
Dichte: 1,15 g/cm³ (20 °C)
Flammpunkt: >250 °C
MDL: MFCD00007850
CAS-Nummer: 101-54-2
EINECS: 202-951-9
UN: 3077

Bestellen

Spezifikation Testergebnisse

Appearance Grey to brown to purple Powder or crystals or flakes or mass and/or chunks
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR