5-Fluoruracil 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2,4-Dihydroxy-5-fluorpyrimidin, Fluoruracil, 5-FU
228440010
228440050
228440250
228440010.EA
28.6
EUR
228440010.
ACRO228440050
ACRO228440250
5-Fluoruracil 99%
5-Fluoruracil
Formel:
C₄H₃FN₂O₂ Molecular Weight: 130.08 g/mol Siedepunkt: 190…200 °C (.1 mmHg) Schmelzpunkt: 278…282 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006018 CAS-Nummer: 51-21-8 EINECS: 200-085-6 UN: 2811 ADR: 6.1 III Merck Index: 13,04208 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % (80°C, 4 hrs) (in vacuum) |
Sulfated ash | ≤0.1 % |
Titration with NaOH | 98.5 to 101.0 % (On dried substance) |
Heavy metals (as Pb) | ≤20 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...