5-Amino-1H-indazol

Lieferant: Thermo Scientific

Synonyme: 5-Amino-1H-indazol, 1H-Indazol-5-ylamin, Indazol-5-ylamin, 1H-Indazol-5-amin, Indazol-5-amin

AC30454.10 AC30454.50
MAYBAC30454.10EA 0 EUR
MAYBAC30454.10 MAYBAC30454.50
5-Amino-1H-indazol
5-Amino-1H-indazole
Formel: C₇H₇N₃
Molecular Weight: 133,15 g/mol
Siedepunkt: 345 °C (1013 hPa)
Schmelzpunkt: 183 °C
MDL: MFCD00037975
CAS-Nummer: 19335-11-6
EINECS: 242-971-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR