5-Amino-2-methylindol

Lieferant: Thermo Scientific

Synonyme: 2-Methyl-5-indolamine, 2-Methylindol-5-ylamine, 5-Amino-2-methyl-1H-indole

AC34875DA AC34875EA
MAYBAC34875DAEA 0 EUR
MAYBAC34875DA MAYBAC34875EA
5-Amino-2-methylindol
5-Amino-2-methylindole
Formel: C₉H₁₀N₂
Molecular Weight: 146.19 g/mol
Schmelzpunkt: 158…159 °C
MDL: MFCD02093426
CAS-Nummer: 7570-49-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR