4-Hydroxy-3,5-dimethylbenzoesäure 98%

Lieferant: Thermo Scientific
Achtung

351760010 351760050 351760250
ACRO351760010EA 39.4 EUR
ACRO351760010 ACRO351760050 ACRO351760250
4-Hydroxy-3,5-dimethylbenzoesäure 98%
4-Hydroxy-3,5-dimethylbenzoesäure
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
Schmelzpunkt: 221…226 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016536
CAS-Nummer: 4919-37-3

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to light beige to light pink Crystalline powder
Infrared spectrum Conforms
Melting point 221 to 226 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR