2-Thiophenethanol 98%
Lieferant: Thermo Scientific
Synonyme:
2-(2-Thienyl)ethanol, 2-(2-Hydroxyethyl)thiophen, Thiophen-2-ethanol
366540250
366541000
ACRO366540250EA
42.8
EUR
ACRO366540250
ACRO366541000
2-Thiophenethanol 98%
2-Thiophenethanol
Formel:
C₆H₈OS Molecular Weight: 128,19 g/mol Siedepunkt: 108 °C (13 mmHg) Dichte: 1,153 g/cm³ Flammpunkt: 101 °C |
MDL:
MFCD00005462 CAS-Nummer: 5402-55-1 EINECS: 226-452-0 |
Spezifikation Testergebnisse
Appearance | Clear light yellow to grey-green to brown Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5490 to 1.5520 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...