5-Brom-2-chlorpyrimidin 96%

Lieferant: Thermo Scientific
Achtung

368482500 368480050
ACRO368482500EA 25.7 EUR
ACRO368482500 ACRO368480050
5-Brom-2-chlorpyrimidin 96%
5-Bromo-2-chloropyrimidine
Formel: C₄H₂BrClN₂
Molecular Weight: 193,43 g/mol
Schmelzpunkt: 78…80 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00483232
CAS-Nummer: 32779-36-5

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR