p-Phenylendiamin Dihydrochlorid 99+%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1,4-Diaminobenzene dihydrochloride, 1,4-Phenylenediammonium dichloride, 1,4-Phenylenediamine dihydrochloride, Benzene-1,4-diamine dihydrochloride
417481000
417480250
ACRO417481000EA
69.9
EUR
ACRO417481000
ACRO417480250
p-Phenylendiamin Dihydrochlorid 99+%
p-Phenylendiamin Dihydrochlorid
Formel:
C₆H₁₀Cl₂N₂ Molecular Weight: 181,06 g/mol Schmelzpunkt: 275 °C Dichte: Min, 1 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013002 CAS-Nummer: 624-18-0 EINECS: 210-834-9 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to yellow to purple-grey Crystalline powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥99.0 % |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...