Palladium(II)nitrat Hydrat (≥39% Pd) ≥99,8% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Palladiumdinitrat Hydrat
11035.04
11035.09
11035.18
11035.04EA
305
EUR
11035.04
11035.09
11035.18
Palladium(II)nitrat Hydrat (≥39% Pd) ≥99,8% (Metall-Basis)
Palladium(II)nitrat Hydrat
Purity: 99.8%
CAS No.: 10102-05-3
Molecular Formula: N2O6Pd.xH2O
Formula Weight: 230.43(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Red to brown
Melting Point: dec.
Solubility: Soluble in water with turbidity, precipitating a brown basic salt. Completely soluble in dilute HNO3
Merck Reference: 14,6992
UN No.: 1477
MDL No.: MFCD00011169
Formel:
N₂O₆Pd·H₂O Molecular Weight: 230.43 g/mol (anhy) Schmelzpunkt: Zersetzung Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011169 CAS-Nummer: 313222-87-6 EINECS: 233-265-8 UN: 1477 ADR: 5.1 III Merck Index: 14,6992 |