Bromcyan

Lieferant: Thermo Scientific
Gefahr

Synonyme: Cyanogenbromid

32731.14 32731.22 32731.06
32731.14EA 58.4 EUR
32731.14 32731.22 32731.06
Bromcyan
Bromcyan

CAS No.: 506-68-3 Molecular Formula: BrCN Formula Weight: 105.93 Physical Form: Crystalline Density: 2.015 Odour: Acrid Boiling Point: 61.4° Merck Reference: 14,2693 UN No.: 1889 MDL No.: MFCD00011597

Formel: CBrN
Molecular Weight: 105,92 g/mol
Siedepunkt: 61,4 °C
Schmelzpunkt: 50…52 °C
Dichte: 2,015
Lagertemperatur: Kühlschrank
MDL: MFCD00011597
CAS-Nummer: 506-68-3
EINECS: 208-051-2
UN: 1889
ADR: 6.1,I
Merck Index: 14,2693

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR