Eisen(II)oxalat Hydrat ≥96%
Lieferant: Thermo Scientific
Achtung
Synonyme: Ethandisäure Eisen(II)salz, Oxalsäure Eisen(II)salz
39261.36 39261.A3
39261.36EA 67.3 EUR
39261.36 39261.A3
Eisen(II)oxalat Hydrat ≥96%
Eisen(II)oxalat Hydrat
Used as a photographic developer, glass tint, decorative glass colorant, and pigment in plastics, paints, and lacquers.
Purity: 96% CAS No.: 516-03-0 Molecular Formula: FeC2O4.xH2O (x≈2) Formula Weight: 143.87(anhy) Physical Form: Powder Density: 2.28 Melting Point: 190° dec. Solubility: Soluble in acids UN No.: 3288 MDL No.: MFCD00012475
Formel: FeC₂O₄·xH₂O
Molecular Weight: 143,87 g/mol (anhy)
Schmelzpunkt: 190 °C (Zersetzung)
Dichte: 2,28
Lagertemperatur: Raumtemperatur
MDL: MFCD00012475
CAS-Nummer: 516-03-0
EINECS: 208-217-4
UN: 3288
ADR: 6.1,III