Dikaliumhexabromoiridat (≥25,2% Ir) ≥99,9% (Metall-Basis)

Lieferant: Thermo Scientific
Achtung

Synonyme: Kaliumhexabromoiridat(IV)

41012.03 41012.06
41012.03EA 204 EUR
41012.03 41012.06
Dikaliumhexabromoiridat (≥25,2% Ir) ≥99,9% (Metall-Basis)
Dikaliumhexabromoiridat

Purity: 99.9% CAS No.: 19121-78-9 Molecular Formula: K2IrBr6 Formula Weight: 749.86 Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Black Odour: Odourless MDL No.: MFCD00054221

Formel: Br₆IrK₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00054221
CAS-Nummer: 19121-78-9
EINECS: 242-827-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR