Kaliumtellurit Hydrat 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
di-Kaliumtellurtrioxid Hydrat, Kaliumtellur(IV)oxid Hydrat
41959.14
41959.22
41959.14EA
151
EUR
41959.14
41959.22
Kaliumtellurit Hydrat 97%
Kaliumtellurit Hydrat
Potassium tellurite, in association with agar is used as a selective medium for growth of clauberg (bacteria). It is used for the selective isolation of staphylococci and corynebacteria, which is used to reduce TeO3 2- to tellurium element.
Purity: 97%
CAS No.: 123333-66-4
Molecular Formula: K2O3Te.H2O
Formula Weight: 276.81 (253.80anhy)
Physical Form: Crystalline
Appearance: White
Melting Point: 465° dec.
UN No.: 3284
MDL No.: MFCD00149923
Formel:
K₂O₃Te·H₂O Molecular Weight: 271,81 g/mol Schmelzpunkt: 465 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149923 CAS-Nummer: 123333-66-4 EINECS: 232-213-1 UN: 3284 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...