3-Hydroxy-5-methylisoxazol ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Hymexazol

44450.06 44450.14 44450.22
44450.06EA 49.9 EUR
44450.06 44450.14 44450.22
3-Hydroxy-5-methylisoxazol ≥97%
3-Hydroxy-5-methylisoxazole

Purity: 97% CAS No.: 10004-44-1 Molecular Formula: C4H5NO2 Formula Weight: 99.09 Physical Form: Powder Solubility: Soluble in alcohol, acetone, THF, chloroform MDL No.: MFCD00144468

Formel: C₄H₅NO₂
Molecular Weight: 99,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00144468
CAS-Nummer: 10004-44-1
EINECS: 233-000-6
Merck Index: 14,4856

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR