4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosan ([2.2.1]Cryptand, Kryptofix® 221) ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Kryptofix® 221, 1,10-Diaza-4,7,13,16,21-pentaoxabicyclo[8.8.5]tricosan, [2.2.1]Cryptand

44504.02 44504.04 44504.77
44504.02EA 218 EUR
44504.02 44504.04 44504.77
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosan ([2.2.1]Cryptand, Kryptofix® 221) ≥97%
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosan ([2.2.1]Cryptand, Kryptofix® 221)

Purity: 97% CAS No.: 31364-42-8 Molecular Formula: C16H32O5N2 Formula Weight: 332.44 Storage Temperature: 2°C to 8°C Physical Form: Liquid Density: 1.11 Refractive Index: 1.5010 Sensitivity: Light Sensitive Appearance: Pale yellow Beilstein Reference: 616444 MDL No.: MFCD00005108

Formel: C₁₆H₃₂N₂O₅
Molecular Weight: 332,44 g/mol
Dichte: 1,11
MDL: MFCD00005108
CAS-Nummer: 31364-42-8
EINECS: 250-592-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR