5-(Chlormethyl)uracil ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-(Chlormethyl)-2,4-dihydroxypyrimidin

44639.01 44639.03 44639.06
44639.01EA 49 EUR
44639.01 44639.03 44639.06
5-(Chlormethyl)uracil ≥97%
5-(Chlormethyl)uracil

Purity: 97% CAS No.: 3590-48-5 Molecular Formula: C5H5ClN2O2 Formula Weight: 160.56 Physical Form: Powder

Formel: C₅H₅ClN₂O₂
Molecular Weight: 160.56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00218445
CAS-Nummer: 3590-48-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR