Nitroprussid-Natrium Dihydrat 99,0-102,0% ACS
Lieferant: Thermo Scientific
Gefahr
Synonyme: Dinatriumpentacyanonitrosylferrat(III) Dihydrat, Natriumnitrosopentacyanoferrat(III) Dihydrat, di-Natriumnitrosylpentacyanoferrat(III) Dihydrat, Natriumnitroprussiat Dihydrat, di-Natriumpentacyanonitrosylferrat(III) Dihydrat
87683.18 87683.30
87683.18EA 67 EUR
87683.18 87683.30
Nitroprussid-Natrium Dihydrat 99,0-102,0% ACS
Nitroprussid-Natrium Dihydrat
Purity: 99.0-102.0% CAS No.: 13755-38-9 Molecular Formula: C5FeN6Na2O.2H2O Formula Weight: 297.95 (261.92anhy) Physical Form: Crystalline Density: 1.72 Appearance: Dark red Odour: Odourless Merck Reference: 14,8649 UN No.: 1588 MDL No.: MFCD00149192
For chromatographic detection of peptides
Formel: Na₂[Fe(CN)₅NO]·2H₂O
Molecular Weight: 297,95 g/mol
Dichte: 1,72
Lagertemperatur: Raumtemperatur
MDL: MFCD00149192
CAS-Nummer: 13755-38-9
EINECS: 238-373-9
UN: 1588
ADR: 6.1,III
Merck Index: 14,8649