Benzothiazol ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Benzo[d]thiazole
A10380.0B
A10380.18
A10380.30
A10380.0BEA
129
EUR
A10380.0B
A10380.18
A10380.30
Benzothiazol ≥97%
Benzothiazole
Purity: 97%
CAS No.: 95-16-9
Molecular Formula: C7H5NS
Formula Weight: 135.19
Flash Point: 107°(224°F)
Refractive Index: 1.6420
Odour: Characteristic
Melting Point: 2°
Boiling Point: 230°
Solubility: Slightly soluble in water. Soluble in alcohol, carbon disulfide
Beilstein Reference: 109468
Merck Reference: 14,1107
UN No.: 2810
MDL No.: MFCD00005775
Formel:
C₇H₅NS Siedepunkt: 230 °C Schmelzpunkt: 2 °C Dichte: 1,241 Flammpunkt: 107 °C (224 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005775 CAS-Nummer: 95-16-9 EINECS: 202-396-2 UN: 2810 ADR: 6.1,III Merck Index: 14,1107 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...