Ethylbromacetat ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Bromessigsäureethylester
A10448.36
A10448.0E
A10448.22
A10448.36EA
93.1
EUR
A10448.36
A10448.0E
A10448.22
Ethylbromacetat ≥98%
Ethylbromacetat
Purity: 98%
CAS No.: 105-36-2
Molecular Formula: C4H7BrO2
Formula Weight: 167.00
Flash Point: 47°(117°F)
Physical Form: Clear liquid
Density: 1.506
Refractive Index: 1.4510
Appearance: Colourless
Melting Point: -38°
Boiling Point: 159°
UN No.: 1603
MDL No.: MFCD00000191
Formel:
BrCH₂COOCH₂CH₃ Molecular Weight: 167 g/mol Siedepunkt: 159 °C Schmelzpunkt: –38 °C Dichte: 1,506 Flammpunkt: 47 °C (117 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000191 CAS-Nummer: 105-36-2 EINECS: 203-290-9 UN: 1603 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...