Diphenylsilandiol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dihydroxydiphenylsilane

A10492.36 A10492.22 A10492.14 AB116343.0100
A10492.36EA 253 EUR
A10492.36 A10492.22 A10492.14 ABCRAB116343.0100
Diphenylsilandiol ≥98%
Diphenylsilandiol

Purity: 98% CAS No.: 947-42-2 Molecular Formula: C12H12O2Si Formula Weight: 216.31 Odour: Odourless Melting Point: ca 150° dec. Beilstein Reference: 2523445 UN No.: 1325 MDL No.: MFCD00002101

Formel: C₁₂H₁₂O₂Si
Molecular Weight: 216.31 g/mol
Siedepunkt: 377
Schmelzpunkt: ∼ 150 °C (Zers.)
Dichte: 1.25
Lagertemperatur: Raumtemperatur
MDL: MFCD00002101
CAS-Nummer: 947-42-2
EINECS: 213-427-4
UN: 1325
ADR: 4.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR