5-Nitrobenzimidazol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Nitro-1H-benzimidazole

A10524.18 A10524.30
A10524.18EA 32.9 EUR
A10524.18 A10524.30
5-Nitrobenzimidazol ≥98%
5-Nitrobenzimidazole

Purity: 98+% CAS No.: 94-52-0 Molecular Formula: C7H5N3O2 Formula Weight: 163.14 Melting Point: 207-211° Beilstein Reference: 7926

Formel: C₇H₅N₃O₂
Molecular Weight: 163.14 g/mol
Schmelzpunkt: 207…211 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005604
CAS-Nummer: 94-52-0
EINECS: 202-341-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR