6-Nitrobenzothiazol ≥98%

Lieferant: Thermo Scientific
Achtung

A10634.06 A10634.18
A10634.06EA 45.3 EUR
A10634.06 A10634.18
6-Nitrobenzothiazol ≥98%
6-Nitrobenzothiazole

Purity: 98+% CAS No.: 2942-06-5 Molecular Formula: C7H4N2O2S Formula Weight: 180.18 Melting Point: 173-177° Beilstein Reference: 7824

Formel: C₇H₄N₂O₂S
Molecular Weight: 180,19 g/mol
Schmelzpunkt: 173…177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014569
CAS-Nummer: 2942-06-5
EINECS: 220-933-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR