4,5-Dimethylthiazol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
A10785.06
A10785.14
A10785.22
A10785.06EA
45.1
EUR
A10785.06
A10785.14
A10785.22
4,5-Dimethylthiazol ≥98%
4,5-Dimethylthiazole
Purity: 98%
CAS No.: 3581-91-7
Molecular Formula: C5H7NS
Formula Weight: 113.18
Flash Point: 51°(123°F)
Refractive Index: 1.5210
Boiling Point: 157-158°
Beilstein Reference: 105694
UN No.: 1993
MDL No.: MFCD00005336
Formel:
C₅H₇NS Molecular Weight: 113,18 g/mol Siedepunkt: 157…158 °C Dichte: 1,070 Flammpunkt: 51 °C (123 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005336 CAS-Nummer: 3581-91-7 EINECS: 222-703-3 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...