2-Amino-4-methylbenzothiazol 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Methylbenzothiazol-2-ylamin
A10822.06
A10822.14
A10822.22
A10822.06EA
28.4
EUR
A10822.06
A10822.14
A10822.22
2-Amino-4-methylbenzothiazol 98%
2-Amino-4-methylbenzothiazole
Purity: 98%
CAS No.: 1477-42-5
Molecular Formula: C8H8N2S
Formula Weight: 164.22
Melting Point: 137-139°
Beilstein Reference: 129346
Formel:
C₈H₈N₂S Molecular Weight: 164.23 g/mol Siedepunkt: 321 Schmelzpunkt: 137…139 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005793 CAS-Nummer: 1477-42-5 EINECS: 216-028-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...