5-Chloruracil ≥98%

Lieferant: Thermo Scientific

Synonyme: 5-Chlor-2,4-dihydroxypyrimidin, Chloruracil

A11084.06 A11084.14
A11084.06EA 43.9 EUR
A11084.06 A11084.14
5-Chloruracil ≥98%
5-Chloruracil

Purity: 98% CAS No.: 1820-81-1 Molecular Formula: C4H3ClN2O2 Formula Weight: 146.53 Melting Point: 300° Beilstein Reference: 127173

Formel: C₄H₃ClN₂O₂
Molecular Weight: 146,53 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006019
CAS-Nummer: 1820-81-1
EINECS: 217-339-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR