Cholsäure ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
3α,7α,12α-Trihydroxycholan-24-säure, 3α,7α,12α-Trihydroxy-5β-cholansäure, 3α,7α,12α-Trihydroxycholansäure, 3α,7α,12α-Trihydroxy-5β-cholan-24-säure
A11257.14
A11257.22
A11257.36
A11257.14EA
34
EUR
A11257.14
A11257.22
A11257.36
Cholsäure ≥98%
Cholsäure
Purity: 98+%
CAS No.: 81-25-4
Molecular Formula: C24H40O5
Formula Weight: 408.58
Melting Point: 199-202°
Beilstein Reference: 2822009
Merck Reference: 14,2203
Optical Rotation: +35 ± 2° (c=0.6 in ethanol)
MDL No.: MFCD00003672
Formel:
C₂₄H₄₀O₅ Molecular Weight: 408.58 g/mol Siedepunkt: 545 Schmelzpunkt: 199…202 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003672 CAS-Nummer: 81-25-4 EINECS: 201-337-8 Merck Index: 13,02223 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...