2-Thenoesäurehydrazid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Thiophencarbonsäurehydrazid, Thiophen-2-carbonsäurehydrazid, Thiophen-2-carbohydrazid

A11270.06 A11270.14
A11270.06EA 47.3 EUR
A11270.06 A11270.14
2-Thenoesäurehydrazid ≥98%
2-Thenoesäurehydrazid

Purity: 98+% CAS No.: 2361-27-5 Molecular Formula: C5H6N2OS Formula Weight: 142.18 Melting Point: 137-140° Beilstein Reference: 114437

Formel: C₅H₆N₂OS
Molecular Weight: 142.18 g/mol
Schmelzpunkt: 137…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005435
CAS-Nummer: 2361-27-5
EINECS: 219-107-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR