2-Methyl-4-pentensäure ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Methylpent-4-enoic acid
A11297.09
A11297.18
A11297.09EA
45.2
EUR
A11297.09
A11297.18
2-Methyl-4-pentensäure ≥98%
2-Methyl-4-pentenoic acid
Purity: 98%
CAS No.: 1575-74-2
Molecular Formula: C6H10O2
Formula Weight: 114.15
Flash Point: 107°(224°F)
Refractive Index: 1.4310
Odour: Characteristic
Boiling Point: 195°
Beilstein Reference: 1720987
UN No.: 3265
MDL No.: MFCD00015570
Formel:
C₆H₁₀O₂ Molecular Weight: 114,14 g/mol Siedepunkt: 195 °C Dichte: 0,950 Flammpunkt: 107 °C (224 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015570 CAS-Nummer: 1575-74-2 EINECS: 216-404-7 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...