2-Amino-4,6-dichlorpyrimidin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4,6-Dichloropyrimidin-2-ylamine
A11330.06
A11330.14
A11330.22
A11330.06EA
46.7
EUR
A11330.06
A11330.14
A11330.22
2-Amino-4,6-dichlorpyrimidin 98%
2-Amino-4,6-dichloropyrimidine
Purity: 98%
CAS No.: 56-05-3
Molecular Formula: C4H3Cl2N3
Formula Weight: 164.00
Melting Point: 218-222°
Beilstein Reference: 118454
Formel:
C₄H₃Cl₂N₃ Molecular Weight: 163,99 g/mol Schmelzpunkt: 218…222 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006090 CAS-Nummer: 56-05-3 EINECS: 200-253-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...