N-Bromphthalimid ≥98%

Lieferant: Thermo Scientific
Achtung

A11601.06 A11601.14 A11601.22
A11601.06EA 30.9 EUR
A11601.06 A11601.14 A11601.22
N-Bromphthalimid ≥98%
N-Bromphthalimid

Purity: 98+% CAS No.: 2439-85-2 Molecular Formula: C8H4BrNO2 Formula Weight: 226.03 Melting Point: 204-208° Beilstein Reference: 131211

Formel: C₈H₄BrNO₂
Molecular Weight: 226,03 g/mol
Schmelzpunkt: 204…208 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005888
CAS-Nummer: 2439-85-2
EINECS: 219-467-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR