Indazol ≥99%

Lieferant: Thermo Scientific

Synonyme: 1H-Indazol

A11665.03 A11665.06 A11665.14
A11665.03EA 24.7 EUR
A11665.03 A11665.06 A11665.14
Indazol ≥99%
Indazol

Purity: 99% CAS No.: 271-44-3 Molecular Formula: C7H6N2 Formula Weight: 118.14 Melting Point: 146-149° Boiling Point: 270° Merck Reference: 14,4936 MDL No.: MFCD00005691

Formel: C₇H₆N₂
Molecular Weight: 118,14 g/mol
Siedepunkt: 270 °C
Schmelzpunkt: 146…149 °C
Dichte: 1,32 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005691
CAS-Nummer: 271-44-3
EINECS: 205-978-4
Merck Index: 14,4936

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR