4'-Methoxybutyrophenon 97%
Lieferant: Thermo Scientific
Synonyme:
1-(4-Methoxyphenyl)butan-1-one
A11737.03
A11737.06
A11737.14
A11737.03EA
27.4
EUR
A11737.03
A11737.06
A11737.14
4'-Methoxybutyrophenon 97%
4'-Methoxybutyrophenon
Purity: 97%
CAS No.: 4160-51-4
Molecular Formula: C11H14O2
Formula Weight: 178.23
Refractive Index: 1.5390
Boiling Point: 124-128°/4mm
MDL No.: MFCD00027138
Formel:
C₁₁H₁₄O₂ Molecular Weight: 178,23 g/mol Siedepunkt: 124…128 °C (4 mmHg) Schmelzpunkt: 26 °C Dichte: 1,0451 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00027138 CAS-Nummer: 4160-51-4 EINECS: 223-995-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...