3-Brom-2-thenoesäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Bromthiophen-2-carbonsäure, 3-Brom-2-thiophencarbonsäure

A11935.03 A11935.06
A11935.03EA 28.1 EUR
A11935.03 A11935.06
3-Brom-2-thenoesäure ≥97%
3-Brom-2-thenoesäure

Purity: 97% CAS No.: 7311-64-0 Molecular Formula: C5H3BrO2S Formula Weight: 207.05 Melting Point: 195-196° Beilstein Reference: 121585

Formel: C₅H₃BrO₂S
Molecular Weight: 207,05 g/mol
Schmelzpunkt: 195…196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052291
CAS-Nummer: 7311-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR