1-Chlor-5-fluor-2-iodbenzol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Chlor-4-fluor-1-iodbenzol

A12010.06 A12010.14 A12010.22
A12010.06EA 26.9 EUR
A12010.06 A12010.14 A12010.22
1-Chlor-5-fluor-2-iodbenzol ≥98%
1-Chlor-5-fluor-2-iodbenzol

Purity: 98% CAS No.: 101335-11-9 Molecular Formula: C6H3ClFI Formula Weight: 256.45 Refractive Index: 1.6010 Boiling Point: 96-98°/15mm MDL No.: MFCD00051774

Formel: C₆H₃ClFI
Molecular Weight: 256,45 g/mol
Siedepunkt: 96…98 °C (15 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00051774
CAS-Nummer: 101335-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR