N-(Chlormethyl)phthalimid ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
N-(Chlormethyl)-1,2-benzoldicarbonsäureimid, N-(Chlormethyl)phthalsäureimid
A12018.14
A12018.22
A12018.14EA
95.3
EUR
A12018.14
A12018.22
N-(Chlormethyl)phthalimid ≥97%
N-(Chlormethyl)phthalimid
Purity: 97%
CAS No.: 17564-64-6
Molecular Formula: C9H6ClNO2
Formula Weight: 195.61
Melting Point: 131-135°
Beilstein Reference: 140942
Formel:
C₉H₆ClNO₂ Molecular Weight: 195,6 g/mol Siedepunkt: 305 °C (1013 hPa) Schmelzpunkt: 131…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005898 CAS-Nummer: 17564-64-6 EINECS: 241-541-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...