N-(2-Bromethyl)phthalimid ≥98%
Lieferant: Thermo Scientific
Synonyme:
N-(2-Bromethyl)phthalsäureimid, N-(2-Bromethyl)-1,2-benzoldicarbonsäureimid
A12164.14
A12164.22
A12164.36
A12164.14EA
33
EUR
A12164.14
A12164.22
A12164.36
N-(2-Bromethyl)phthalimid ≥98%
N-(2-Bromethyl)phthalimid
Purity: 98+%
CAS No.: 574-98-1
Molecular Formula: C10H8BrNO2
Formula Weight: 254.09
Melting Point: 80-84°
Beilstein Reference: 148736
Formel:
C₁₀H₈BrNO₂ Molecular Weight: 254,08 g/mol Siedepunkt: 318 °C (1013 hPa) Schmelzpunkt: 80…84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005902 CAS-Nummer: 574-98-1 EINECS: 209-379-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...