5-Amino-3-methylisoxazol 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Methylisoxazol-5-ylamine, 3-Methylisoxazol-5-amine

A12446.06 A12446.03 A12446.14
A12446.06EA 108 EUR
A12446.06 A12446.03 A12446.14
5-Amino-3-methylisoxazol 98%
5-Amino-3-methylisoxazole

Purity: 98+% CAS No.: 14678-02-5 Molecular Formula: C4H6N2O Formula Weight: 98.11 Melting Point: 81-85° Beilstein Reference: 107906

Formel: C₄H₆N₂O
Molecular Weight: 98.1 g/mol
Siedepunkt: 194
Schmelzpunkt: 81…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003151
CAS-Nummer: 14678-02-5
EINECS: 238-719-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR