5-Nitrouracil ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,4-Dihydroxy-5-nitropyrimidin, Nitrouracil

A12448.14 A12448.22 A12448.36
A12448.14EA 45.5 EUR
A12448.14 A12448.22 A12448.36
5-Nitrouracil ≥98%
5-Nitrouracil

Purity: 98+% CAS No.: 611-08-5 Molecular Formula: C4H3N3O4 Formula Weight: 157.09 Melting Point: ca 300° Beilstein Reference: 10410

Formel: C₄H₃N₃O₄
Molecular Weight: 157,09 g/mol
Schmelzpunkt: ∼ 300 °C
Dichte: 1,807 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006021
CAS-Nummer: 611-08-5
EINECS: 210-250-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR