2-Thenoesäure ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: Thiophen-2-carbonsäure, 2-Thiophencarbonsäure

A12514.14 A12514.22 A12514.36
A12514.14EA 25.7 EUR
A12514.14 A12514.22 A12514.36
2-Thenoesäure ≥99%
2-Thenoesäure

Purity: 99% CAS No.: 527-72-0 Molecular Formula: C5H4O2S Formula Weight: 128.15 Melting Point: 126-130° Boiling Point: 259-261° Merck Reference: 14,9354 MDL No.: MFCD00005437

Formel: C₅H₄O₂S
Molecular Weight: 128,15 g/mol
Siedepunkt: 259…261 °C
Schmelzpunkt: 126…130 °C
Dichte: 1,42
Lagertemperatur: Raumtemperatur
MDL: MFCD00005437
CAS-Nummer: 527-72-0
EINECS: 208-423-4
Merck Index: 14,9354

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR