Phthalimid ≥99%
Lieferant: Thermo Scientific
Synonyme:
1,2-Benzoldicarbonsäureimid, Phthalsäureimid
A12584.0C
A12584.0E
A12584.36
A12584.0CEA
304
EUR
A12584.0C
A12584.0E
A12584.36
Phthalimid ≥99%
Phthalimid
Purity: 99%
CAS No.: 85-41-6
Molecular Formula: C8H5NO2
Formula Weight: 147.14
Flash Point: 165°(329°F)
Odour: Odourless
pH: 3.8
Melting Point: 232-236°
Boiling Point: 366°
Solubility: Slightly soluble in water. Soluble in aqueous alkali hydroxides
Beilstein Reference: 118522
Merck Reference: 14,7373
MDL No.: MFCD00005881
Formel:
C₈H₅NO₂ Molecular Weight: 147,13 g/mol Siedepunkt: 366 °C Schmelzpunkt: 232…236 °C Dichte: 1,21 Flammpunkt: 165 °C (329 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005881 CAS-Nummer: 85-41-6 EINECS: 201-603-3 Merck Index: 14,7373 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...