6-Aminobenzothiazol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Benzothiazol-6-ylamin
A12601.03
A12601.06
A12601.03EA
52
EUR
A12601.03
A12601.06
6-Aminobenzothiazol ≥98%
6-Aminobenzothiazole
Purity: 98+%
CAS No.: 533-30-2
Molecular Formula: C7H6N2S
Formula Weight: 150.20
Melting Point: 86-90°
Beilstein Reference: 116381
Merck Reference: 14,425
MDL No.: MFCD00015461
Formel:
C₇H₆N₂S Molecular Weight: 150,2 g/mol Schmelzpunkt: 86…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015461 CAS-Nummer: 533-30-2 EINECS: 208-559-4 Merck Index: 14,425 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...