trans-2-Pentensäure ≥90% remainder other isomers
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(E)-Pent-2-enoic acid, 2-Pentenoic acid
A12689.09
A12689.18
A12689.09EA
52
EUR
A12689.09
A12689.18
trans-2-Pentensäure ≥90% remainder other isomers
trans-2-Pentenoic acid
Purity: 90+%
CAS No.: 13991-37-2
Molecular Formula: C5H8O2
Formula Weight: 100.12
Flash Point: 102°(215°F)
Refractive Index: 1.4515
Odour: Pungent
Melting Point: 8-10°
Boiling Point: 200-203°
Beilstein Reference: 1720312
UN No.: 3265
MDL No.: MFCD00002704
Formel:
C₅H₈O₂ Molecular Weight: 100,12 g/mol Siedepunkt: 200…203 °C Schmelzpunkt: 8…10 °C Dichte: 0,992 Flammpunkt: 102 °C (215 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002704 CAS-Nummer: 13991-37-2 EINECS: 237-791-9 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...