α,α,α-Trifluor-3,4-diaminotoluol 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-(Trifluormethyl)benzol-1,2-diamin, 4-(Trifluormethyl)-1,2-phenylendiamin, 1,2-Diamino-4-(trifluormethyl)benzol, 3,4-Diaminobenzotrifluorid, 4-(Trifluormethyl)-o-phenylendiamin

A12749.03 A12749.06
A12749.03EA 38.3 EUR
A12749.03 A12749.06
α,α,α-Trifluor-3,4-diaminotoluol 98%
α,α,α-Trifluor-3,4-diaminotoluol

Purity: 98% CAS No.: 368-71-8 Molecular Formula: C7H7F3N2 Formula Weight: 176.14 Melting Point: 58-62° Beilstein Reference: 909258 UN No.: 2811 MDL No.: MFCD00042456

Formel: C₇H₇F₃N₂
Molecular Weight: 176.14
Siedepunkt: 101…102 °C (7 mmHg)
Schmelzpunkt: 58…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042456
CAS-Nummer: 368-71-8
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR