5-Fluor-2-indolcarbonsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Fluoro-1H-indole-2-carboxylic acid

A12794.03 A12794.06 A12794.14
A12794.03EA 34.9 EUR
A12794.03 A12794.06 A12794.14
5-Fluor-2-indolcarbonsäure ≥98%
5-Fluoroindole-2-carboxylic acid

Purity: 98+% CAS No.: 399-76-8 Molecular Formula: C9H6FNO2 Formula Weight: 179.15 Melting Point: ca 259° dec. Beilstein Reference: 153217

Formel: C₉H₆FNO₂
Molecular Weight: 179,15 g/mol
Schmelzpunkt: ∼ 259 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005612
CAS-Nummer: 399-76-8
EINECS: 206-919-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR