2-Pyrrolcarbaldehyd ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Pyrrol-2-carboxaldehyd, 1H-Pyrrol-2-carbaldehyd, Pyrrol-2-carbaldehyd, 1H-Pyrrol-2-carboxaldehyd
A12838.09
A12838.14
A12838.22
A12838.09EA
56.2
EUR
A12838.09
A12838.14
A12838.22
2-Pyrrolcarbaldehyd ≥99%
2-Pyrrolcarbaldehyd
Purity: 99%
CAS No.: 1003-29-8
Molecular Formula: C5H5NO
Formula Weight: 95.10
Storage Temperature: 2°C to 8°C
Flash Point: 106°(222°F)
Melting Point: 40-46°
Boiling Point: 217-219°
Beilstein Reference: 105745
MDL No.: MFCD00005217
Formel:
C₅H₅NO Molecular Weight: 95,1 g/mol Siedepunkt: 217…219 °C Schmelzpunkt: 40…46 °C Flammpunkt: 106 °C (222 °F) |
MDL:
MFCD00005217 CAS-Nummer: 1003-29-8 EINECS: 213-705-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...