(4-Hydroxyphenoxy)essigsäure ≥98%

Lieferant: Thermo Fisher Scientific

A12940.06 A12940.14 A12940.22
A12940.06EA 49.7 EUR
A12940.06 A12940.14 A12940.22
(4-Hydroxyphenoxy)essigsäure ≥98%
(4-Hydroxyphenoxy)essigsäure

Purity: 98+% CAS No.: 1878-84-8 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 153-158° Beilstein Reference: 1952779

Formel: (C₆H₄(OH))OCH₂COOH
Molecular Weight: 168.15 g/mol
MDL: MFCD00014362
CAS-Nummer: 1878-84-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR